Learn More
Sartorius Picus™ NxT Electronic Pipette, 8 Channels, Trade-In
Designed to ease user's workload and to protect from repetitive strain injury (RSI)
Products 4
Catalog Number Mfr. No. Color Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Color Volume (Metric) Price Quantity & Availability  
14-557-827PR
View Documents
Sartorius
LH745321PR
Gray 0.2 to 10 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
14-557-828PR
View Documents
Sartorius
LH745341PR
Yellow 5 to 120 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
14-557-829PR
View Documents
Sartorius
LH745361PR
Orange 10 to 300 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
14-557-830PR
View Documents
Sartorius
LH745391PR
Purple 50 to 1200 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
Laboratory work requires repetition of similar pipetting steps that may cause strain on hand and arm, and thus may compromise results. To minimize this risk, a highly ergonomic electronic pipette should be used for minimum effort and strain. Sartorius Picus™ NxT are the most sophisticated and ergonomic electronic compact and light weight pipettes.
  • Password protection for stored programs
  • Very last liquid droplets dispensed by repeated blow-out for maximum recovery
  • Features one-touch operating-button and feather-light electronic tip ejection
  • Includes plate tracker for microwell plate dispensing
  • Electronic piston control and brake for precise pipetting
  • Safe-cone filters to prevent contamination
  • Adjustment wheel for volume setting and navigation
  • Range of pipetting modes for diluting and titrating to serial dispensing
  • Intuitive user interface in a choice of language options: English, French, German, Russian and Chinese
  • Certificate of accredited 3-point calibration
  • RFID-enabled to support asset management
  • Volume range: 0.2μL to 1200μL
Applications:
PCR and other DNA/RNA techniques, ELISA, Protein analysis and Cell culture
Offer Details
After purchasing this trade-in program item please follow these steps to complete your trade in:
1. Complete Sartorius redemption form. (Contact Fisher Customer Service to obtain the form.)
2. Sartorius will email a link to a prepaid UPS shipping label, shipping instructions, and decontamination form.
3. Visit UPS site, print the shipping label, pack pipette, and ship to Sartorius.
Note: Any brand pipette, in any condition can be traded in. Pipettes must be free of contamination and returned together with the signed Decontamination Form.
Certifications
CE Marked
Warranty and Services
2 year