Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MTA2, Synthetic Peptide, Invitrogen™

Synthetic Peptide

Manufacturer:  Invitrogen PEP0102

Catalog No. PEP0102


Add to cart

Description

Description

PID Synthetic Peptide, PEP-0102, from Invitrogen.

This peptide corresponds to 17 amino acids of human PID.PEP-0102 can be used as a blocking peptide with polyclonal antibody PA5-19978.
Specifications

Specifications

Synthetic Peptide
purified
50μg
Blocking Assay, Control
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
Human
RUO
0.02% sodium azide
MTA2
Blocking Assay, Control
9219
Human
200μg/mL
Liquid
N
PBS
-20°C
Human MTA2
O94776
Peptide
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit