Learn More
PIERCAN USA CSM Long Gloves
Manufacturer:  PIERCAN USA INC 6Y15328.5
 View more versions of this product
Catalog No. 17-010-100
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Ambidextrous
6 in.
UV rays, ozone, reducing Acids, diluted Acids, Alkalis and Salts
260°F
15mil
32 in.
CSM (Chlorosulfonated Polyethylene)
150mm
8.5
126°C
White
812mm