Learn More
PIERCAN USA High Performance Butyl Short Isolater Gloves
Manufacturer:  PIERCAN USA INC BHP156A
 View more versions of this product
Catalog No. 17-010-403
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Ambidextrous
Ketones, Acids, Alkalis and Salts
280°F
15mil
13 in.
Butyl
6
137°C
Black
330mm