Learn More
PIERCAN USA Neoprene Long Gloves, 6 in. Port Size
Neoprene gloves feature high mechanical properties and are generally resistant to hydrogen peroxide, oil and grease. Available in ambidextrous or left/right specific hand styles.
$4100.00 - $5080.00
Specifications
Color Black
Length (English) 32 in.
Length (Metric) 81.2 cm
Material Neoprene
Port Size (English) 6 in.
View More Specs
Products 21
Catalog Number Mfr. No. Size Hand Thickness (English) Thickness (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Size Hand Thickness (English) Thickness (Metric) Price Quantity & Availability  
17-010-242
View Documents
PIERCAN USA
6N15328A
8 Ambidextrous 15 mil 0.38 mm
Case of 10 for $4,100.00
Add to cart
17-010-244
View Documents
PIERCAN USA
6N15329A
9 Ambidextrous 15 mil 0.38 mm
Case of 10 for $4,100.00
Add to cart
17-010-246
View Documents
PIERCAN USA
6N153210A
10 Ambidextrous 15 mil 0.38 mm
Case of 10 for $4,100.00
Add to cart
17-010-247
View Documents
PIERCAN USA
6N153211A
11 Ambidextrous 15 mil 0.38 mm
Case of 10 for $4,100.00
Add to cart
17-010-253
View Documents
PIERCAN USA
6N24328A
8 Ambidextrous 24 mil 0.60 mm
Case of 10 for $4,660.00
Add to cart
17-010-255
View Documents
PIERCAN USA
6N24329A
9 Ambidextrous 24 mil 0.60 mm
Case of 10 for $4,660.00
Add to cart
17-010-257
View Documents
PIERCAN USA
6N243210A
10 Ambidextrous 24 mil 0.60 mm
Case of 10 for $4,660.00
Add to cart
17-010-258
View Documents
PIERCAN USA
6N243211A
11 Ambidextrous 24 mil 0.60 mm
Case of 10 for $4,660.00
Add to cart
17-010-264
View Documents
PIERCAN USA
6N30328A
8 Ambidextrous 30 mil 0.76 mm
Case of 10 for $5,080.00
Add to cart
17-010-266
View Documents
PIERCAN USA
6N30329A
9 Ambidextrous 30 mil 0.76 mm
Case of 10 for $5,080.00
Add to cart
17-010-268
View Documents
PIERCAN USA
6N303210A
10 Ambidextrous 30 mil 0.76 mm
Case of 10 for $5,080.00
Add to cart
17-010-269
View Documents
PIERCAN USA
6N303211A
11 Ambidextrous 30 mil 0.76 mm
Case of 10 for $5,080.00
Add to cart
17-010-241
View Documents
PIERCAN USA
6N15328
8 Specific 15 mil 0.38 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
17-010-243
View Documents
PIERCAN USA
6N15329
9 Specific 15 mil 0.38 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
17-010-245
View Documents
PIERCAN USA
6N153210
10 Specific 15 mil 0.38 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
17-010-252
View Documents
PIERCAN USA
6N24328
8 Specific 24 mil 0.60 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
17-010-254
View Documents
PIERCAN USA
6N24329
9 Specific 24 mil 0.60 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
17-010-256
View Documents
PIERCAN USA
6N243210
10 Specific 24 mil 0.60 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
17-010-263
View Documents
PIERCAN USA
6N30328
8 Specific 30 mil 0.76 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
17-010-265
View Documents
PIERCAN USA
6N30329
9 Specific 30 mil 0.76 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
17-010-267
View Documents
PIERCAN USA
6N303210
10 Specific 30 mil 0.76 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Neoprene provides chemical resistance with reduced risk of sensitization
  • High mechanical properties
  • Offers dexterity and flexibility
  • Resistant to ozone and UV
  • Protects up to 260degreeF
  • Ambidextrous
  • Ideal industries include pharmaceutical, nuclear, biotech, microelectronic, defense and universities
  • Compatible with 6 in. port
Specifications
Specifications
Black
81.2 cm
6 in.
32 in.
Neoprene
15 cm