Learn More
PIERCAN USA Polyurethane/Chlorosulfonated Polyethylene Long Gloves
Manufacturer:  PIERCAN USA INC 6UY20329.75
 View more versions of this product
Catalog No. 17-010-152
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Left/Right
6 in.
High mechanical properties, puncture and tear resistance, highest performance with in situ leak detectors, high resistance to UV Rays/Ozone, natural aging, very resistant to reducing Acids, diluted Acids, Alkalis and Salts
160°F
20 mil
32 in.
CSM (Chlorosulfonated Polyethylene),Polyurethane
150mm
9.75
71°C
White
812mm