Learn More
Thermo Scientific™ Pierce™ 0.1% Trifluoroacetic Acid (v/v) in Acetonitrile, LC-MS Grade
An LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.
Supplier: Thermo Scientific™ 85177
Description
Thermo Scientific™ Pierce 0.1% Trifluoroacetic Acid in Acetonitrile is an LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.
Additional Information and Online Ordering »
Highlights:
Validated
– tested more than 20 ways to ensure maximal LC/MS sensitivity
Purified
– processed, filtered and sealed to extend LC/MS column life
Convenient
– packaged to eliminate variability and reduce handling
Specifications
LC-MS | |
4 x 1 L | |
Liquid | |
Pierce™ |
Trifluoroacetic Acid (TFA) | |
Store in a cool, dry, and well-ventilated area. Protect from direct sunlight. | |
Solvent Mixture, Ion Pairing Reagent | |
HPLC, Mass Spectrometry |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.