Learn More
Thermo Scientific™ Pierce™ ABTS Substrates
Ready-to-use, water-soluble peroxidase substrate tablets that yield a measurable green end-product for use in ELISA methods.
Products 1
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
PI34026
View Documents
Thermo Scientific™
34026
Pierce ABTS Tablets This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Thermo Scientific™ Pierce ABTS Substrates are tablets and a ready-to-use solution of a water-soluble peroxidase substrate that yields a measurable green end product for use in ELISA methods.
ABTS (2,2'-Azinobis [3-ethylbenzothiazoline-6-sulfonic acid]-diammonium salt) is a water-soluble HRP substrate that yields a green end product upon reaction with peroxidase. The green product has two major absorbance peaks, 410nm and 650nm. ABTS is less sensitive than OPD and TMB in ELISA applications. It is less readily oxidized, and its color development is slower (approximately 20 minutes). This may be advantageous if unacceptable background results from the use of the OPD or TMB substrates due to higher sensitivities.
Highlights:
  • Excellent choice when maximum sensitivities are not required
  • Less readily oxidized and color development is slower (approximately 20 minutes) than OPD and TMB