Learn More
Thermo Scientific™ Pierce™ Top 12 Abundant Protein Depletion Spin Columns
Decrease the abundant albumin and antibody components of human plasma samples in preparation for mass spectrometry or 1D and 2D gel electrophoresis.
Manufacturer: Thermo Scientific™ 85165
Description
Decrease the abundant albumin and antibody components of human plasma samples in preparation for mass spectrometry or 1D and 2D gel electrophoresis with Thermo Scientific™ Pierce Abundant Protein Depletion Spin Columns.
Additional Information and Online Ordering »
Highlights:
Optimized – resin in spin columns is scaled and optimized for treating 10μL (600μg) of human plasma samples for mass spectrometry and/or 1D and 2D electrophoresis
Efficient – kits remove >90% of IgG and >95% of albumin, plus other abundant proteins (Top 12)
Fast – process samples in 40 to 60 minutes (depending on resin)
Economical – cost effective spin columns are priced for single use
Reduce variability – One time use prevents abundant protein carryover and experimental variability
Two resin options – choose the system appropriate for your need: 2 or 12 protein depletion columns
Top 2 Spin Columns Remove:
- Albumin
- IgG
Top 12 Spin Columns Remove:
- Albumin
- IgG
- α1-Acid Glycoprotein
- α1-Antitrypsin
- α2-Macroglobulin
- Apolipoprotein A-I
- Apolipoprotein A-II
- Fibrinogen
- Haptoglobin
- IgA
- IgM
- Transferrin
Specifications
24 Columns | |
Mass Spectrometry | |
Plasma, Serum | |
Microcentrifuge, Mass Spectrometer | |
Pierce™ |
Sample Enrichment, Sample Clean Up | |
Protein | |
Pierce Top 12 Abundant Protein Depletion Spin Columns | |
Spin Column |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.