Learn More
Thermo Scientific™ Pierce™ Top 12 Abundant Protein Depletion Spin Columns
GSA_VA
Decrease the abundant albumin and antibody components of human plasma samples in preparation for mass spectrometry or 1D and 2D gel electrophoresis.
Products 2
Catalog Number Mfr. No. Description dimQuantity Price Quantity & Availability  
Catalog Number Mfr. No. Description dimQuantity Price Quantity & Availability  
PI85165
GSA_VA
View Documents Promotion Details
Thermo Scientific™
85165
Pierce Top 12 Abundant Protein Depletion Spin Columns This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
PI85164
View Documents Promotion Details
Thermo Scientific™
85164
Pierce Top 12 Abundant Protein Depletion Spin Columns This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Decrease the abundant albumin and antibody components of human plasma samples in preparation for mass spectrometry or 1D and 2D gel electrophoresis with Thermo Scientific™ Pierce Abundant Protein Depletion Spin Columns.
Additional Information and Online Ordering »
The Pierce Top 2 Abundant Protein Depletion Spin Columns use highly specific immobilized anti-HSA and anti-IgG antibodies to remove human serum albumin (HSA) and all major subclasses of gamma globulin (IgG) from serum, plasma or spinal fluids. Similarly, the Pierce Top 12 Abundant Protein Depletion Spin Columns are designed to remove HSA, IgG and 10 other high-abundance proteins from human serum or plasma. The resins are provided in an economical and convenient spin column specifically designed one step processing and for single use.
Highlights:
  • Optimized – resin in spin columns is scaled and optimized for treating 10μL (600μg) of human plasma samples for mass spectrometry and/or 1D and 2D electrophoresis
  • Efficient – kits remove >90% of IgG and >95% of albumin, plus other abundant proteins (Top 12)
  • Fast – process samples in 40 to 60 minutes (depending on resin)
  • Economical – cost effective spin columns are priced for single use
  • Reduce variability – One time use prevents abundant protein carryover and experimental variability
  • Two resin options – choose the system appropriate for your need: 2 or 12 protein depletion columns
Top 2 Spin Columns Remove:
  • Albumin
  • IgG
Top 12 Spin Columns Remove:
  • Albumin
  • IgG
  • α1-Acid Glycoprotein
  • α1-Antitrypsin
  • α2-Macroglobulin
  • Apolipoprotein A-I
  • Apolipoprotein A-II
  • Fibrinogen
  • Haptoglobin
  • IgA
  • IgM
  • Transferrin
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.