Learn More
Thermo Scientific™ Pierce™ Avidin Agarose
Description
Thermo Scientific™ Pierce Avidin Agarose is high-quality beaded agarose resin for biotin affinity procedures.
Additional Information and Online Ordering
Avidin is a tetrameric glycoprotein rich in tryptophan that is present in chicken egg white and the tissues of birds, and avidin is highly resistant to denaturation by acids or proteolytic enzymes. Avidin Agarose Resin consists of purified avidin that has been immobilized to crosslinked 6% beaded agarose. The resulting affinity support is leak-resistant, stable at pH from 2 to 11, and ideal for gravity-flow and low-speed centrifugation applications.
Highlights:
- Immobilized ligand is purified avidin from chicken egg whites
- Support is crosslinked 6% beaded agarose resin (agarose beads)
- Supplied as a stable aqueous slurry or pre-packed columns
- Binds free biotin and biotinylated molecules with high affinity
Recommended for:
Binding biotinylated anti-transferrin from serum; Binding biotinyl peptides and elution with an SDS/urea solution; Hybridization of biotinylated RNA to its complementary DNA and binding to immobilized avidin, with subsequent elution of the single-stranded DNA; Purification of double-stranded DNA

Specifications
Specifications
Capacity (Metric) | 20 μg Biotin Binding per mL Resin |
Content And Storage | Store at 4°C. Do not freeze. |
Product Line | Pierce™ |
Quantity | 5 mL |
Target | Biotin-Tagged Molecule |
Type | Agarose |
Stationary Phase | Avidin |
Format | Bottle |
Column Type | Affinity, Agarose Resin |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.