Learn More
Thermo Scientific™ BS3 (bis(sulfosuccinimidyl)suberate)
Crosslink amines to amines with BS3; composed of sulfo-NHS-ester groups separated by an 11.4A, water-soluble spacer arm that is useful for cell-surface protein crosslinking.
Manufacturer:  Thermo Scientific™ 21585
 View more versions of this product
Catalog No. PI21585
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Thermo Scientific™ Pierce BS3 (Sulfo-DSS) is bis(sulfosuccinimidyl)suberate, an amine-to-amine crosslinker that is homobifunctional, water-soluble, non-cleavable and membrane impermeable.
Highlights:
  • Crosslink cell-surface proteins before cell lysis and immunoprecipitation
  • Identify receptor-ligand interactions by chemical crosslinking
  • “Fix” protein interactions to allow identification of weak or transient protein interactions
  • Identify near-neighbor protein interactions
  • Protein crosslinking to create bioconjugates via single-step reactions
  • Immobilize proteins onto amine-coated surfaces
Specifications
Specifications
Amine-to-Amine
Pierce BS3 (Sulfo-DSS), No-Weigh Format
No
No
28mL of typical 1X (1mM) reaction solution (total), or 8 separate uses
Yes
No
Bis(sulfosuccinimidyl) suberate; solid
572.43
8 x 2mg
11.4Å