Learn More
Thermo Scientific™ Pierce™Dimethylsulfoxide (DMSO), Sequencing Grade
A sequencing-grade DMSO preparation with high purity and consistent absorptivity properties for use in HPLC and mass spectrometry methods.
Supplier:  Thermo Scientific™ TS20688
Catalog No. TS20688
Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative.
Description
Description
Thermo Scientific™ Pierce™ Dimethylsulfoxide (DMSO) is a sequencing-grade preparation with high purity for use in traditional amino acid analyzers and other high-volume applications.
This sequencing-grade dimethylsulfoxide is >99.5% pure DMSO which is an excellent choice for protein and peptide applications that require the water-miscible solvent. The purity test ensures that preparation is essentially devoid of amines. This DMSO is packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps for ultimate protection.
Specifications
Specifications
Flammable
Room Temperature
950 mL
Pierce™
Dimethylsulfoxide (DMSO)
Liquid
Clear, colorless liquid of dimethylsulfoxide (at least 99.5% pure; low water)
LC/MS
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.