Learn More
Thermo Scientific™ DSS (disuccinimidyl suberate), No-Weigh™ Format
Crosslink amines to amines with disuccinimidyl suberate composed of NHS-ester groups with a membrane-permeable, mid-length spacer for intracellular protein crosslinking.
$88.90 - $182.80
Specifications
Solubility DMF, DMSO
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PI21655
View Documents Promotion Details
Thermo Scientific™
21655
50 mg
Pack for $90.90
Add to cart
PI21555
View Documents Promotion Details
Thermo Scientific™
21555
1 g
Pack for $182.80
Add to cart
PIA39267
View Documents Promotion Details
Thermo Scientific™
A39267
10 x 2 mg
Each for $88.90
Add to cart
Description
Description
Thermo Scientific™ Pierce DSS, also called disuccinimidyl suberate, is noncleavable and membrane permeable crosslinker that contains an amine-reactive N-hydroxysuccinimide (NHS) ester at each end of an 8-carbon spacer arm.
Highlights:
  • Chemical crosslinking of intracellular proteins prior to cell lysis and immunoprecipitation
  • “Fix” protein interactions to allow identification of weak or transient protein interactions
  • Protein crosslinking to create bioconjugates via single-step reactions
  • Crosslink oriented antibodies to Protein A or G agarose
  • Immobilize proteins onto amine-coated surfaces
Specifications
Specifications
DMF, DMSO