Learn More
Thermo Scientific™ Pierce™ DTT (Dithiothreitol), No-Weigh™ Format
Reduce disulfides in biological buffers and sample loading buffers with pure, crystalline dithiothreitol (DTT), the most popular protein disulfide reducing agent.
$177.00
Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
PIA39255
View Documents
Thermo Scientific™
A39255
Each for $177.00
Add to cart
PI20291
View Documents
Thermo Scientific™
20291
Pierce DTT, No-Weigh Format This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Thermo Scientific™ Pierce DTT (Dithiothreitol) is offered as 7.7mg x 48-tube microplates for convenient preparation of 100µL of 0.5M stocks for addition to protein samples for effective reduction of disulfide bonds.
Highlights:
  • A water-soluble reagent that reduces disulfide bonds
  • Maintains mono-thiols completely in the reduced state and reduces disulfide bonds quantitatively
  • Specific and sensitive assay for disulfides using DTT with Ellman's Reagent