Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ ECL Western Blotting Substrate: PROMO

Save 25% Off

Value-priced, entry-level peroxidase substrate for enhanced chemiluminescence (ECL) that directly replaces costlier products without the need to re-optimize conditions.

Specifications

Conjugate HRP
Description Pierce ECL Western Blotting Substrate
Formulation Proprietary luminol and peroxide solutions
View More Specs

Includes: Detection Reagent 1 and 2

Products 2
Catalog Number Mfr. No. Quantity Kit Contents Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Kit Contents Price Quantity & Availability  
PI32209SPCL
View Documents
Thermo Scientific™
0032209SPCL
3 x 250 mL Kit Luminol/Enhancer, 125mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
PI32106SPCL
View Documents
Thermo Scientific™
0032106SPCL
500 mL Kit Luminol/Enhancer, 250mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Pierce ECL Western Blotting Substrate provides reliability and performance equivalent to other standard enhanced chemilumescent (ECL) substrates for detection of horseradish peroxidase (HRP) enzyme activity, yet costs much less. Because the luminol and peroxide reagent-formulations are identical to other commercially available substrate products, one can switch to Pierce ECL without needing to optimize probing conditions or incubation protocols. Simply switch to Pierce ECL and save a bundle.

Highlights:

  • ECL substrate – an enhanced chemiluminescent substrate for detection of horseradish peroxidase (HRP) activity from antibodies and other Western blot probes

  • About half the cost of other ECL Substrates – low overhead and a commitment to customer value enables Thermo Fisher Scientific to make this product available to you at a significantly lower price than other companies (these claims are based on the 2010 U.S. list prices)

  • No optimization required – switching to Pierce ECL substrate from other entry-level ECL substrates does not require optimization or protocol changes

  • A name you can can rely on – count on the strong quality assurance, technical support and reputation behind Thermo Scientific Pierce Protein Research Products

Specifications

Specifications

HRP
Pierce ECL Western Blotting Substrate
Proprietary luminol and peroxide solutions
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit