Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Factor Xa

Protease (serine endopeptidase for prothrombin); cleaves arginine in the sequence Ile-Glu-Gly-Arg; useful for His-tagged fusion protein engineering.

Manufacturer:  Thermo Scientific™ 32520

 View more versions of this product

Catalog No. PI32520

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Thermo Scientific™ Pierce Factor Xa is a serine endopeptidase composed of two disulfide-linked subunits, whose specificity makes it particular useful in cleaving 6xHis tags from engineered His-tagged fusion proteins.

Cleaves after the arginine residue in its preferred cleavage site Ile-(Glu or Asp)-Gly-Arg and will occasionally cleave at other basic residues. However, it will not cleave at a site followed by proline or arginine. Often used to remove fusion tags, such as the common histidine tag, from expressed proteins.

Highlights:

  • Extremely specific proteolytic activity - useful in the genetic engineering of proteins
  • Cleaves after the last amino acid in the sequence Ile-Glu-Gly-Arg
  • Activity is >130U/mg using a synthetic Ile-Glu-Gly-Arg peptide with p-nitroanilide derivatized arginine as the substrate; each unit of enzyme cleaves 1mol/minute of substrate at 37°C in 5mM MES, 0.5M NaCl, 1mM benzamidine at pH 6 with 1% BSA

Additional Features:

  • Source: Bovine Plasma
  • Activity: 125units/mg
  • Supplied: ≥5mM MES, pH 6, 0.5M NaCl
  • Protein Concentration: 1-2mg/mL

Ordering Information:

Supplied as frozen liquid.

Specifications

Specifications

250 μg
At least 1mg/mL enzyme in MES-buffered saline pH 6 with 1mM benzamidine
At least 250 units of activity/mg
Pierce Factor Xa
No
Safety and Handling

Safety and Handling

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit