Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ GST (glutathione-S-transferase)

Pure, lyophilized glutathione S-transferase (GST) protein from Schistosoma japonicum for use in recombinant fusion protein research methods and assays.

Specifications

Description Pierce GST
Formulation GST (>/=100U/mg protein) in clear, colorless, viscous liquid, free of foreign material
Sufficient For Controls and tests for GST fusion protein purification
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PI20239
View Documents Promotion Details
Thermo Scientific™
20239
10 x 1 mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Thermo Scientific™ GST (Glutathione S-Transferase) is pure, lyophilized glutathione S-transferase for use in a variety of protein research procedures.

This pure GST from Schistosoma japonicum is useful in a variety of applications involving GST-tagged fusion proteins and glutathione affinity purification. The expression of foreign proteins in E. coli using a glutathione S-transferase (GST) fusion system is a method of choice for studying the function of particular proteins, for purifying interacting factors and for generating antibodies against fused proteins. Analyses involving GST-tagged proteins usually require or benefit from the use of pure GST as a control or standard.

Highlights:

  • Purified from Schistosoma japonicum
  • Useful as a molecular weight marker in gel analysis of GST-tagged proteins
  • Suitable as a standard or control in ELISA and pull-down assays involving GST-tagged proteins
  • Ideal for testing tag-specific cross-reactivity of antibodies in ELISA
Specifications

Specifications

Pierce GST
GST (>/=100U/mg protein) in clear, colorless, viscous liquid, free of foreign material
Controls and tests for GST fusion protein purification
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit