Learn More
Thermo Scientific™ Pierce™ HRV 3C Protease Solution Kit (2 units/μL)
Remove fusion tags from proteins with the HRV 3C cleavage sequence with this recombinant cysteine protease that is dual tagged for easy removal from the sample after cleavage.
$383.00 - $3280.00
Specifications
Content And Storage | Store at -20°C. |
---|---|
Product Line | Pierce™ |
Form | Kit |
Product Type | Protease |
Description
Remove fusion tags from proteins with the HRV 3C cleavage sequence with Thermo Scientific™ Pierce Human Rhinovirus (HRV) 3C Protease, a recombinant cysteine protease that is dual tagged for easy removal from the sample after cleavage.
Additional Information and Online Ordering »
Highlights:
Effective – HRV 3C is designed to remove purification tags from your protein of interest
High activity at 4°
C – activity is ≥2000U/mg control protein; one unit will cleave 0.1mg control protein at 4°C after 16h
Flexible – dual-tagged enzyme is compatible with various columns and buffers
Includes:
Kit contains protease, cleavage buffer and control protein.
Recommended for:
Removal of purification tags from fusion proteins
Specifications
Store at -20°C. | |
Pierce™ | |
Kit | |
Protease |
For Research Use Only. Not for use in diagnostic procedures.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.