Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Iodination Beads

Easily and gently label (iodinate) proteins and peptides with iodine-125 using reagent-coated polystyrene beads that can be removed following reaction.

Specifications

Description Pierce Iodination Beads
Formulation Polystyrene beads (3mm dia.) coated with iodination reagent
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PI28666
View Documents
Thermo Scientific™
28666
250 beads This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Thermo Scientific™ Pierce Iodination Beads are 3mm diameter polystyrene beads that are coated with an oxidizing reagent, which provides efficient activation of iodine-125 for protein or peptide iodination procedures.

Highlights:

  • Derivatized, uniform, nonporous polystyrene beads
  • Radioiodide incorporation as high as 99%; labeled protein recovery >90%
  • Functions over a broad pH and temperature range
  • Fast and easy to use: iodinations complete in 2 to 15 minutes
  • Reaction stopped by simply removing beads from reaction mixture with tweezers or pasteur pipet; no reducing agent necessary to terminate reaction

Additional Features:

  • Iodinates in the presence of azides, detergents, urea and high salt
  • Allows efficient iodination of cell membrane surface proteins
  • More gentle method for iodination than soluble chloramine-T because there is no contact between the protein and the immobilized oxidizing agent1
  • Highly reproducible iodinations
  • Can be used to quantitatively iodinate histidine at pH 8.22
Specifications

Specifications

Pierce Iodination Beads
Polystyrene beads (3mm dia.) coated with iodination reagent
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit