Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Pierce™ Iodination Beads
Easily and gently label (iodinate) proteins and peptides with iodine-125 using reagent-coated polystyrene beads that can be removed following reaction.
Supplier: Thermo Scientific™ 28665
Description
Thermo Scientific™ Pierce Iodination Beads are 3mm diameter polystyrene beads that are coated with an oxidizing reagent, which provides efficient activation of iodine-125 for protein or peptide iodination procedures.
Highlights:
- Derivatized, uniform, nonporous polystyrene beads
- Radioiodide incorporation as high as 99%; labeled protein recovery >90%
- Functions over a broad pH and temperature range
- Fast and easy to use: iodinations complete in 2 to 15 minutes
- Reaction stopped by simply removing beads from reaction mixture with tweezers or pasteur pipet; no reducing agent necessary to terminate reaction
Additional Features:
- Iodinates in the presence of azides, detergents, urea and high salt
- Allows efficient iodination of cell membrane surface proteins
- More gentle method for iodination than soluble chloramine-T because there is no contact between the protein and the immobilized oxidizing agent1
- Highly reproducible iodinations
- Can be used to quantitatively iodinate histidine at pH 8.22
Specifications
Iodination Bead | |
Upon receipt store product desiccated at 4°C. Avoid exposing beads to moisture or reducing agents. Product is shipped at ambient temperature. | |
Iodine-125 |
50 Beads | |
Pierce™ | |
Radioisotope |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction