Learn More
Thermo Scientific™ Pierce™ LDH Cytotoxicity Assay Kit
Quantitatively measure lactose dehydrogenase released into the media from damaged cells as a biomarker for cellular cytotoxicity and cytolysis using a colorimetric assay.
$148.10
Specifications
Description Pierce LDH Cytotoxicity Assay Kit
Formulation Five-component kit
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PI88953
View Documents
Thermo Scientific™
88953
200 Reaction Kit
Each for $148.10
Add to cart
PI88954
View Documents
Thermo Scientific™
88954
1000 Reaction Kit This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Quantitatively measure lactose dehydrogenase (LDH) released into the media from damaged cells as a biomarker for cellular cytotoxicity and cytolysis using a colorimetric assay with Thermo Scientific™ Pierce LDH Cytotoxicity Assay Kit.
Additional Information and Online Ordering »
The Pierce LDH Cytotoxicity Assay Kit measures extracellular LDH in culture media using an enzymatic reaction that results in a red formazan product which can be measured spectrophotometrically. Lactate dehydrogenase (LDH) is a cytosolic enzyme that is is an indicator of cellular toxicity. The assay is ideal for high-throughput screening and provides a safe alternative to radioactive cytotoxicity assays.
Highlights:
  • Convenient – add–mix–read assay format for adherent and suspension cells
  • Colorimetric – quantitatively measures LDH release by formation of colored product
  • Robust – uses stable LDH enzyme activity as a cytotoxic marker
  • Flexible – ideal for high–throughput screening
  • Non–radioactive – safe alternative to 51Cr–release cytotoxicity assays
Includes:
Kit contains substrate mix, assay buffer, 10X lysis buffer, stop solution, and LDH positive control
Requires:
Microplate reader capable of reading absorbance at 490nm and 680nm, flat-bottom clear 96–well plate compatible with spectrophotometer, multichannel pipette
Recommended for:
  • Measure in vitro cytotoxicity mediated by chemicals, immune cells, or siRNA or microRNA
  • Detect cytolysis in bioreactor or 3D tissue engineering applications
Specifications
Specifications
Pierce LDH Cytotoxicity Assay Kit
Five-component kit