Learn More
Thermo Scientific™ Pierce™ Trypsin Protease, MS Grade
GSA_VA
Ensure maximum activity and stability in proteomic applications with this chemically modified, highly purified porcine trypsin.
$85.80 - $380.00
Products 2
Catalog Number Mfr. No. Description dimQuantity Price Quantity & Availability  
Catalog Number Mfr. No. Description dimQuantity Price Quantity & Availability  
PI90058
GSA_VA
View Documents Promotion Details
Thermo Scientific™
90058
Pierce Trypsin Protease, MS Grade
Pack of 5 for $380.00
Add to cart
PI90057
GSA_VA
View Documents Promotion Details
Thermo Scientific™
90057
Pierce Trypsin Protease, MS Grade
Pack of 5 for $85.80
Add to cart
Description
Description
Thermo Scientific™ Pierce MS Grade Trypsin Protease is highly purified porcine trypsin that has been chemically modified for maximum activity and stability in proteomic applications.
Additional Information and Online Ordering »
Pierce Trypsin Protease, MS Grade, is a serine protease derived from porcine pancreatic extracts. The enzyme is TPCK-treated to eliminate chymotryptic activity and methylated to improve stability during protein digestion. This mass spectrometry-grade, modified trypsin is then repurified, lyophilized in glass vials and packaged in convenient 5 x 20μg, 5 x 100μg, and 1mg fill sizes.
Highlights:
  • Exceptional selectivity – cleaves at the carboxyl side of lysine and arginine residues with greater than 95% specificity
  • High purity – no detectable chymotrypsin activity
  • Enhanced stability – modified and lyophilized for reduced autolytic activity
  • Economical – available in larger fill sizes that are more cost effective
Recommended for:
  • In-gel digestion of proteins from 1–D or 2–D gels
  • In-solution tryptic digestion of proteins
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.