Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Mini Tablets

Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Mini Tablets broad-spectrum protease and pghospahatase inhibitor formulation provides complete protection from dephosphorylation and proteolytic degradation.

$254.28

Specifications

Sufficient For 1 tablet is suffcient for 10mL
Includes 1 vial of inhibitor tablets
For Use With (Application) protein purification, western blotting
Storage Requirements 2 to 8°C
View More Specs

Alert:

For Research Use Only. Not for use in diagnostic procedures.


Products 2
Catalog Number Mfr. No. Quantity Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Product Type Price Quantity & Availability  
PIA32959 View Documents Thermo Scientific™
A32959
20 tablets Each for $254.28
Add to cart
 
PIA32961 View Documents Thermo Scientific™
A32961
20 tablets Each for $254.28
Add to cart
 
Description

Description

The Pierce Protease and Phosphatase Inhibitor Mini Tablets contain aprotinin, bestatin, E-64, EDTA, and leupeptin to provide protection from degradation by serine-proteases, cysteine-proteases, aspartic acid-proteases. To prevent the dephosphorylation activity of serine/threonine and tyrosine phosphatases, each tablet also contains sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate. Pierce Protease and Phosphatase Inhibitor Tablets are stable at 4°C for up to 12 months, and each tablet is sufficient for 10mL of solution.

Highlights:
  • Improved formulation—tablets dissolve quickly into a clear solution
  • Economical – more cost-effective than other commercially available formulations for equivalent volumes
  • Convenient&mdsash;ready-to-use, fully disclosed, broad-spectrum formulations
  • Compatible—use directly with Thermo Scientific™ Pierce™ BCA Assays
* EDTA is present only in Product No. A32959.
Specifications

Specifications

1 tablet is suffcient for 10mL
1 vial of inhibitor tablets
protein purification, western blotting
2 to 8°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit