Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Protein Concentrators PES, 50K MWCO, 2–6 mL

Thermo Scientific Pierce Protein Concentrators PES are disposable ultrafiltration centrifugal devices with a polyethersulfone (PES) membrane for concentration, desalting, and buffer exchange of biological samples.

$96.30 - $190.50

Specifications

Format Spin Column (15 mL conical)
Product Line Pierce™
Shipping Condition Room Temperature
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PI88538
View Documents
Thermo Scientific™
88538
10 concentrators
Each for $96.30
Add to cart
 
PI88539
View Documents
Thermo Scientific™
88539
24 Concentrators
Each for $190.50
Add to cart
 
Description

Description

This product has a molecular weight cutoff (MWCO) of 50K and can be used for processing sample volumes ranging from 2 to 6 mL.

Features of Pierce Protein Concentrators PES:

  • Rapid protein concentration—sample concentration of 10- to 30-fold can be achieved in 5 to 30 minutes for 10K MWCO (time may vary for a product with a different MWCO)
  • Excellent protein recovery—typical protein recovery is >90%
  • Versatility—perform sample concentration, diafiltration, or buffer-exchange of biological samples
  • Compatibility—may be used in both fixed angle and swing bucket rotors, collect and recover sample without invert spinning

For easy identification, the MWCO value is etched on the side of each device. To assist with sample addition and recovery, a clear window with graduations is located on the side of each device enabling visual determination of the retentate volume. The vertical design of the membrane allows high flux rates, low nonspecific binding, and negligible protein aggregation on the membrane, and it offers reliable and consistent results.

Pierce Protein Concentrators PES contain a membrane rated for retaining molecules with a molecular weight at least 2-fold greater than the MWCO rating of the PES membrane within the device. Reduced recovery may occur when using a concentrator with molecules smaller than the recommended mass. Protein recovery will vary depending on the specific protein in the sample and its starting concentration. To achieve >90% recovery of protein, the minimum protein sample concentration should be 0.1 mg/mL.

Specifications

Specifications

Spin Column (15 mL conical)
Pierce™
Room Temperature
Safety and Handling

Safety and Handling

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit