Learn More
Thermo Scientific™ R-Phycoerythrin
Create fluorescent probes (em 480, 545, 565; ex 578) using this red-fluorescent, multi-subunit protein that can be crosslinked or conjugated to other molecules.
Supplier: Thermo Scientific™ 46185
Description
Thermo Scientific™ R-Phycoerythrin is a red-fluorescent, multi-subunit protein that can be crosslinked or conjugated to other molecules to make fluorescent probes.
The large phycobiliprotein R-phycoerythrin is an excellent dye for applications such as flow cytometry and immunofluorescence that require high sensitivity and/or multicolor detection. R-phycoerythrin makes an ideal probe because it has an intense excitation and emission at long wavelengths where there is little interference from other biological compounds, and because it has a large Stokes shift and high quantum yield. In addition, phycoerythrin is water-soluble and has multiple potential sites for labeling.
Highlights:
- Molecular Weight: 240,000
- Excitation Wavelength: 480, 545, 565nm
- Emission Wavelength: 578nm
References:
Oi, V.T., et al. (1982). Fluorescent phycobiliprotein conjugates for analyses of cells and molecules. J. Cell Biol. 93, 981-986.
Kronick, M.N. and Grossman, P.D. (1983). Immunoassay techniques with fluorescent phycobiliprotein conjugates. Clin. Chem. 29, 1582-1586.
Parks, D.R. and Herzenberg, L.A. (1984). Fluorescence-activated cell sorting: theory, experimental optimization, and applications in lymphoid-cell biology. Meth. Enzymol. 108, 197-241.
Kronick, M.N. (1986). The use of phycobiliproteins as fluorescent labels in immunoassay. J. Immunol. Meth. 92, 1-13.
White, J.C. and Stryer, L. (1987). Photostability studies of phycobiliprotein fluorescent labels. Anal. Biochem. 161, 442-452.
Festin, R., et al. (1987). Detection of triple antibody-binding lymphocytes in standard single laser flow cytometry using colloidal gold, fluorescein and phycoerythrin as labels. J. Immunol. Meth. 101, 23-28.
Specifications
2 mg | |
Water | |
Amine, Carboxylic Acid | |
PE and APC, Fluorescent Protein | |
578 |
R-Phycoerythrin | |
Upon receipt store at 4°C protected from light. Do not freeze. Product shipped with an ice pack. | |
PE and APC | |
Pierce™ | |
480, 545, 565 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.