Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptavidin, PE, Invitrogen™

Protein

Manufacturer:  Invitrogen 21627

Catalog No. PI21627


Add to cart

Description

Description

Streptavidin Protein, RPE conjugate, 21627, from Invitrogen., 21627, from Invitrogen. Species Reactivity: ; Applications: ELISA, Immunohistochemistry, Western Blot.

Streptavidin is biotin-binding protein that was originally isolated from Streptomyces avidinii. In contrast to avidin, streptavidin has no carbohydrate and has a mildly acidic pI of 5. Thermo Scientific Streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI. Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin. Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation. Phycobiliprotein Concentration: 0.5mg/mL (determined by absorption = 82.0 at 566 nm for a 1% solution for only those R-PE molecules to which at least one molecule of active streptavidin is bound) Phycoerythrin Source: Seaweed Restoration and Storage: The expiration date is six months from date of reconstitution. Store freeze-dried product at 4°C until reconstitution. Restore with 1.0 ml distilled water. Centrifuge product if it is not completely clear after reconstitution. To judge clarity, draw product into a pasteur pipette. Store at 4°C. [Do not freeze]. After further dilution of the reconstituted streptavidin, do not use for more than one day. Country of origin: USA
Specifications

Specifications

ELISA,Immunohistochemistry,Western Blot
Lyophilized
Streptavidin
1mL
RUO
Tag
Tag
ELISA, Immunohistochemistry, Western Blot
PBS with 15 mg/mL BSA and 0.05% sodium azide
purified
4°C
Streptavidin
PE
Safety and Handling

Safety and Handling

ShelfLife : 12 Months

SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit