Learn More
Thermo Scientific™ Pierce™ TiO2 Magnetic Phosphopeptide Enrichment Kit
Purify phosphorylated peptides from complex biological samples using titanium dioxide-coated magnetic beads; ideal for mass spectrometry preparation.
Specifications
Formulation Multi-component kit with TiO2 magnetic beads
Kit Contents Binding Buffer, 100mL
Support Type Magnetic Bead
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PI88811
View Documents
Thermo Scientific™
88811
96 rxn kit This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Thermo Scientific™ Pierce Titanium Dioxide Magnetic Phosphopeptide Enrichment Kit is for isolating phosphopeptides from complex biological samples using titanium dioxide-coated magnetic beads.
The Magnetic Phosphopeptide Enrichment Kit is for isolating phosphopeptides from complex biological samples using titanium dioxide-coated magnetic beads. The titanium dioxide (TiO2) ligand on these magnetic particles selectively binds peptides containing phosphorylated serine (Ser), tyrosine (Tyr) or threonine (Thr), enabling selective phosphopeptide enrichment from protease-digested samples. The isolated phosphopeptides are compatible for analysis downstream by mass spectrometry. The binding, wash and elution buffers supplied in the kits ensure that maximum specificity and yield.
The high-performance iron oxide superparamagnetic particles are validated and optimized for use with high-throughput magnetic platforms, such as the Thermo Scientific KingFisher™ 96 and KingFisher Flex Instruments. The beads also enable premium performance for simple benchtop applications using an appropriate magnetic stand.
Highlights:
  • Complete MS-compatible kits – include ready-to-use binding, wash and elution buffers that are optimized for phosphopeptide enrichment and downstream analysis by MALDI and ESI mass spectrometry
  • Optimized for HTS – procedure validated for processing 1 to 96 samples at a time; complete entire assay in about 15 minutes using a KingFisher Flex Instrument
  • Stable affinity ligand – titanium dioxide is specially coated as a film on the magnetic particles
  • Selective – affinity system is selective for phosphorylated Ser, Tyr and Thr; exhibits minimal non-specific binding to acidic residues
  • Sensitive – affinity provides more than 1000 times greater sensitivity than traditional IMAC technologies; enables enrichment and MS-measurement of less than 100 fmol of phosphoprotein
Includes:
Pierce TiO2 Magnetic Beads; Binding, wash and elution buffers; 96-well PCR plates for automated processing
Requires:
Thermo Scientific KingFisher 96 or KingFisher Flex Instrument or other magnetic separator device
Specifications
Specifications
Multi-component kit with TiO2 magnetic beads
Binding Buffer, 100mL
Magnetic Bead