Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TNBSA Solution (2,4,6-trinitrobenzene sulfonic acid) (5% w/v)

Label primary amines (peptides or amino acids) for colorimetric detection (Amax 335nm) using this 5% solution of trinitrobenzene sulfonic acid in methanol.

Supplier:  Thermo Scientific™ 28997

Catalog No. PI28997

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Thermo Scientific™ Pierce TNBSA is a 5% solution of trinitrobenzene sulfonic acid in methanol that reacts with primary amines (peptides or amino acids) to yield a soluble colored product, a property useful for various assay methods.

Highlights:

  • Couples with primary amines, sulfhydryls and hydrazides in aqueous solution at pH 8, without undesirable side reactions
  • Excellent for solution or solid phase analysis
  • Suitable for qualitative and quantitative estimation of biomolecules
  • λmax = 335nm
  • Reacts readily with primary amino groups of amino acids in aqueous format at pH 8 to form yellow adducts
  • No colored derivatives are formed with secondary amino acids proline and hydroxyproline
  • Colored derivatives are monitored at 345nm and have extinction coefficients in range of 1-1.5 x 104
Specifications

Specifications

TNBSA Solution
Storage: Upon receipt store at -20°C. Product is shipped at ambient temperature.
Colorimetric
Liquid
Amine
Other Label or Dye
100 mL
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.