Learn More
PIG3 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 148281AP
Catalog No. 50-172-9758
Add to cart
Description
Description
The protein encoded by this gene is similar to oxidoreductases, which are enzymes involved in cellular responses to oxidative stresses and irradiation. This gene is induced by the tumor suppressor p53 and is thought to be involved in p53-mediated cell death. It contains a p53 consensus binding site in its promoter region and a downstream pentanucleotide microsatellite sequence. P53 has been shown to transcriptionally activate this gene by interacting with the downstream pentanucleotide microsatellite sequence. The microsatellite is polymorphic, with a varying number of pentanucleotide repeats directly correlated with the extent of transcriptional activation by p53. It has been suggested that the microsatellite polymorphism may be associated with differential susceptibility to cancer. At least two transcript variants encoding the same protein have been found for this gene.
Specifications
Specifications
PIG3
0.45 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q53FA7
p53 induced gene 3 protein, PIG3, Quinone oxidoreductase PIG3, TP53I3
Rabbit
IgG
150 μL
-20°C
Primary
9540
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
TP53I3
Liquid
TP53I3
PIG3 Fusion Protein Ag6618
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human