Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIGA Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548364

Catalog No. PA548364

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

This gene encodes a protein required for synthesis of N-acetylglucosaminyl phosphatidylinositol (GlcNAc-PI), the first intermediate in the biosynthetic pathway of GPI anchor. The GPI anchor is a glycolipid found on many blood cells and which serves to anchor proteins to the cell surface. Paroxysmal nocturnal hemoglobinuria, an acquired hematologic disorder, has been shown to result from mutations in this gene. Alternate splice variants have been characterized.
Specifications

Specifications

PIGA
Polyclonal
PBS with 0.09% sodium azide
P37287
Phosphatidylinositol N-acetylglucosaminyltransferase subunit A, GlcNAc-PI synthesis protein, Phosphatidylinositol-glycan biosynthesis class A protein, PIG-A, PIGA
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5277
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PIGA
Liquid
PIGA
KLH conjugated synthetic peptide between 455-484 amino acids from the C-terminal region of human PIGA.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit