Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIGB Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549281

Catalog No. PA549281

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Mannosyltransferase involved in glycosylphosphatidylinositol-anchor biosynthesis. Transfers the third alpha-1,2-mannose to Man2-GlcN-acyl-PI during GPI precursor assembly.
Specifications

Specifications

PIGB
Polyclonal
PBS with 0.09% sodium azide
Q92521
GPI mannosyltransferase 3, 241-, GPI mannosyltransferase III, GPI-MT-III, Phosphatidylinositol-glycan biosynthesis class B protein, PIG-B, PIGB
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9488
Western Blot
Unconjugated
PIGB
Liquid
PIGB
KLH conjugated synthetic peptide between 28-56 amino acids from the N-terminal region of human PIGB.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit