Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIGC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | PIGC |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179871
|
Novus Biologicals
NBP179871 |
100 μL |
Each of 1 for $482.50
|
|
|||||
Description
PIGC Polyclonal specifically detects PIGC in Human samples. It is validated for Western Blot.Specifications
PIGC | |
Polyclonal | |
Rabbit | |
NP_002633 | |
5279 | |
Synthetic peptide directed towards the C terminal of human PIGCThe immunogen for this antibody is PIGC. Peptide sequence AVGAVLFALLLMSISCLCPFYLIRLQLFKENIHGPWDEAEIKEDLSRFLS. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
EC 2.4.1.198, GPI2class C protein, MGC2049, phosphatidylinositol glycan anchor biosynthesis, class C, phosphatidylinositol glycan, class C | |
PIGC | |
IgG | |
33 kDa |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title