Description
Large adult pigeon specimens provide an in-depth view of several traits that all birds share:
- Skin with feathers
- Wings
- Two legs
- Four-chambered heart
- Horny beak
Order Info
INJECTION KEY
Plain: Circulatory system not injected with colors
Single: Arteries injected with red latex
Double: Arteries red, veins blue
Triple: Arteries red, veins blue and hepatic portal veins yellow
Specifications
Pigeon | |
Fisher-Free™ | |
Plain | |
10 Pk. |