Learn More
Pigeons
SCI_ED
Columba sp.
$146.10
Specifications
Type Pigeon
View More Specs
Products 5
Catalog Number Mfr. No. Injection Type Preservative Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Injection Type Preservative Quantity Price Quantity & Availability  
S2402
SCI_ED
View Documents
Ranaco Corporation
138.1008.1
Plain Fisher-Free™ 1 Pk.
Pack for $18.65
N/A
Due to product restrictions, please to purchase or view availability for this product.
S2402VS
SCI_ED
View Documents
Ranaco Corporation
138.1008.10
Plain Fisher-Free™ 10 Pk.
Pack of 10 for $146.10
Add to cart
S2403S
SCI_ED
View Documents
Ranaco Corporation
138.1020.1
Double Fisher-Free™ 1 Pk.
Pack for $19.46
N/A
Due to product restrictions, please to purchase or view availability for this product.
S2404S
SCI_ED
View Documents
N/A Triple Formaldehyde 1 Pk. Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
S2400S
SCI_ED
View Documents
N/A Single Formaldehyde 1 Pk. Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Large adult pigeon specimens provide an in-depth view of several traits that all birds share:
  • Skin with feathers
  • Wings
  • Two legs
  • Four-chambered heart
  • Horny beak
Order Info
INJECTION KEY
Plain: Circulatory system not injected with colors
Single: Arteries injected with red latex
Double: Arteries red, veins blue
Triple: Arteries red, veins blue and hepatic portal veins yellow
Specifications
Specifications
Pigeon