Learn More
Invitrogen™ PIGL Monoclonal Antibody (2B6)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA520744
Catalog No. PIMA520744
Only null left
Add to Cart
Description
Description
Peptide Sequence: GHSNHIALYA AVRALHSEGK LPKGCSVLTL QSVNVLRKYI SLLDLPLSLL HTQDVLFVLN SKEVAQAKKA MSCHRSQLLW FRRLYIIFSR YMRINSLSFL.
This gene encodes an enzyme that catalyzes the second step of glycosylphosphatidylinositol (GPI) biosynthesis, which is the de-N-acetylation of N-acetylglucosaminylphosphatidylinositol (GlcNAc-PI). Study of a similar rat enzyme suggests that this protein localizes to the endoplasmic reticulum.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PIGL
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q9Y2B2
PIGL
PIGL (NP_004269, 153 a.a. ~ 252 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2b κ
ELISA
2B6
Unconjugated
PIGL
CHIME; N-acetylglucosaminylphosphatidylinositol deacetylase; N-acetylglucosaminyl-phosphatidylinositol de-N-acetylase; phosphatidylinositol glycan anchor biosynthesis class L; phosphatidylinositol glycan anchor biosynthesis, class L; phosphatidylinositol glycan, class L; phosphatidylinositol-glycan biosynthesis class L protein; PIGL; PIG-L
Mouse
Affinity chromatography
RUO
9487
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.