Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIGM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561510
Description
Immunogen sequence: QDRTLHVRYT DIDYQVFTDA ARFVTEGRSP YLRATYRYT Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
PIGM encodes a transmembrane protein that is located in the endoplasmic reticulum and is involved in GPI-anchor biosynthesis. The glycosylphosphatidylinositol (GPI)-anchor is a glycolipid which contains three mannose molecules in its core backbone. The GPI-anchor is found on many blood cells and serves to anchor proteins to the cell surface. PIGM encodes a mannosyltransferase, GPI-MT-I, that transfers the first mannose to GPI on the lumenal side of the endoplasmic reticulum.
Specifications
PIGM | |
Polyclonal | |
Unconjugated | |
PIGM | |
4933437L05Rik; AI644422; AV302080; C920011G20Rik; dol-P-Man dependent GPI mannosyltransferase; DPM:GlcN-(acyl-)PI mannosyltransferase; GPI mannosyltransferase 1; GPI mannosyltransferase I; GPI-MT-I; phosphatidylinositol glycan anchor biosynthesis class M; phosphatidylinositol glycan anchor biosynthesis, class M; phosphatidylinositol glycan, class M; phosphatidylinositol-glycan biosynthesis class M protein; Pigm; PIG-M; PIG-M mannosyltransferase; PIG-M mRNA for mannosyltransferase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
93183 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H3S5 | |
PIGM | |
Recombinant protein corresponding to Human PIGM. Recombinant protein control fragment (Product #RP-99782). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction