Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ pIgR Antibody Pair [HRP]
Mouse Monoclonal Antibody Pair
$482.50 - $1183.00
Specifications
Assay Range | 62.5 pg/mL |
---|---|
Assay Sensitivity | 62.5-4000 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Product Type | Antibody Pairs |
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human pIgR.Specifications
62.5 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Human | |
FLJ22667, hepatocellular carcinoma associated protein TB6, Hepatocellular carcinoma-associated protein TB6, MGC125361, MGC125362, PIgR, Poly-Ig receptor, polymeric immunoglobulin receptor | |
PIGR | |
Immunology | |
pIgR |
62.5-4000 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human pIgR. | |
Sandwich ELISA | |
5284 | |
Mouse | |
RUO | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 1.00 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 4000 pg/mL is recommended. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title