Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIGW Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | PIGW |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP16001720
![]() |
Novus Biologicals
NBP16001720UL |
20 μL |
Each for $206.00
|
|
|||||
NBP160017
![]() |
Novus Biologicals
NBP160017 |
100 μL |
Each for $487.50
|
|
|||||
Description
PIGW Polyclonal specifically detects PIGW in Human, Mouse samples. It is validated for Western Blot.Specifications
PIGW | |
Polyclonal | |
Rabbit | |
Q7Z7B1 | |
284098 | |
IgG | |
57 kDa |
Western Blot | |
Unconjugated | |
RUO | |
class W, EC 2.3, FLJ37433, phosphatidylinositol glycan anchor biosynthesis, class W, PIG-W | |
PIGW | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title