Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIH1D2 Mouse anti-Human, Clone: OTI1A2, liquid, TrueMAB™
Mouse Monoclonal Antibody
Supplier: OriGene TA502362
Description
The function of this protein remains unknown.Specifications
PIH1D2 | |
Monoclonal | |
0.6 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8WWB5 | |
PIH1D2 | |
Full length human recombit protein of human PIH1D2 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI1A2 | |
Unconjugated | |
PIH1D2 | |
PIH1 domain containing 2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
120379 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction