Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIK3AP1 Mouse anti-Human, Clone: OTI7A11, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA501812
Description
PIK3AP1 (Phosphoinositide-3-Kinase Adaptor Protein 1) is a Protein Coding gene. Diseases associated with PIK3AP1 include Hemangioma Of Lung. Among its related pathways are HIV Life Cycle and PI3K/AKT activation. Gene Ontology (GO) annotations related to this gene include identical protein binding and phosphatidylinositol 3-kinase regulatory subunit binding. An important paralog of this gene is BANK1.Specifications
PIK3AP1 | |
Monoclonal | |
0.98 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q6ZUJ8 | |
BCAP | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
118788 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI7A11 | |
Unconjugated | |
PIK3AP1 | |
Liquid | |
PIK3AP1 | |
Full length human recombit protein of human PIK3AP1 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title