Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIK3C2B Monoclonal Antibody (OTI2G3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA526242
Description
PIK3C2B Monoclonal antibody specifically detects PIK3C2B in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
PIK3C2B | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00750 | |
PIK3C2B | |
Full length human recombinant protein of PIK3C2B produced in HEK293T cell | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI2G3 | |
Unconjugated | |
PIK3C2B | |
C2-PI3K; C330011J12Rik; Gm199; phosphatidylinositol 3-kinase C2 domain-containing beta polypeptide; phosphatidylinositol 4-phosphate 3-kinase C2 domain-containing subunit beta; phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit beta; phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 beta; phosphatidylinositol-4-phosphate 3-kinase, catalytic subunit type 2 beta; phosphoinositide 3-kinase-C2-beta; phosphoinositide-3-kinase, class 2, beta polypeptide; pi3kc2b; PI3KC2beta; PI3K-C2beta; PI3K-C2-beta; PIK3C2B; PTDINS-3-kinase C2 beta; ptdIns-3-kinase C2 subunit beta | |
Mouse | |
Affinity Chromatography | |
RUO | |
5287 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction