Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIK3CA Mouse anti-Human, Mouse, Rat, Biotin, Clone: OTI6D9, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273406B
Description
PIK3CA Monoclonal specifically detects PIK3CA in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
PIK3CA | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
PIK3CA | |
Recombinant protein fragment corresponding to amino acids 303-631 of human PIK3CA (NP_006209) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Immunohistochemistry | |
OTI6D9 | |
Immunohistochemistry | |
EC 2.7.1, EC 2.7.1.153, MGC142161, MGC142163, p110-alpha, Phosphatidylinositol 3-Kinase catalytic subunit alpha, phosphatidylinositol 3-kinase, catalytic, 110-KD, alpha, phosphatidylinositol 3-kinase, catalytic, alpha polypeptide, Phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha isoform, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit, alpha isoform, phosphoinositide-3-kinase, catalytic, alpha polypeptide, PI3K, PI3K-alpha, PI3-kinase p110 subunit alpha, PI3-kinase subunit alpha, PtdIns-3-kinase p110, PtdIns-3-kinase subunit alpha, PtdIns-3-kinase subunit p110-alpha | |
Mouse | |
Affinity Purified | |
Breast Cancer, Cancer, Diabetes Research, mTOR Pathway, Oncogenes | |
5290.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title