Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIK3CA Rabbit anti-Human, Polyclonal, Abnova™

Rabbit Polyclonal Antibody

Manufacturer:  Abnova Corporation PAB30573

Catalog No. 89-032-914

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Rabbit polyclonal antibody raised against partial recombinant human PIK3CA.

  • High specificity
  • High reactivity
Specifications

Specifications

PIK3CA
Unconjugated
Immunofluorescence (1 - 4 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50 - 1:200) The optimal working dilution should be determined by the end user.
P42336
MGC142161|MGC142163|PI3K|p110-alpha
Rabbit
IgG
100 uL
Store at 4°C for short term storage. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Immunofluorescence, Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against partial recombinant human PIK3CA.
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide).
Liquid
PIK3CA
Recombinant protein corresponding to amino acids 453-573 of human PIK3CA.
Antigen affinity purification
RUO
Primary
5290
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)