Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIK3R3 Rabbit anti-Human, Clone: SR44-06, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP267329

Catalog No. NBP267329


Add to cart

Description

Description

PIK3R3 Monoclonal antibody specifically detects PIK3R3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

PIK3R3
SR44-06
Unconjugated
Purified
100% homology to SWISS-PROT Q92569, DKFZp686P05226, FLJ41892, p55, Phosphatidylinositol 3-kinase 55 kDa regulatory subunit gamma, phosphoinositide-3-kinase, regulatory subunit 3 (gamma), phosphoinositide-3-kinase, regulatory subunit 3 (p55, gamma), phosphoinositide-3-kinase, regulatory subunit, polypeptide 3 (p55, gamma), PI3K regulatory subunit gamma, PI3-kinase subunit p55-gamma, regulatory subunit, polypeptide 3 (p55, gamma)
PIK3R3
Recombinant Protein
Monoclonal
Protein A purified
Signal Transduction
Human,Mouse,Rat
Western Blot,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin)
1 mg/mL
Western Blot 1:1000-1:10000 Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:200-1:500
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide
8503
Rabbit
IgG
Primary
100μL
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit