Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIK3R5 Monoclonal Antibody (OTI2A3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Supplier: OriGene TA505794
Description
Receptor-regulated class I phosphoinositide 3-kinases (PI3Ks) phosphorylate the membrane lipid phosphatidylinositol 4,5-bisphosphate (PtdIns(4,5)P2) to PtdIns(3,4,5)P3, which in turn recruits and activates cytosolic effectors involved in proliferation, survival, or chemotaxis. PIK3R5 is a PI3K regulatory subunit.Specifications
PIK3R5 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8WYR1 | |
PIK3R5 | |
Full length human recombit protein of human PIK3R5 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI2A3 | |
Unconjugated | |
PIK3R5 | |
F730038I15Rik, FOAP-2, P101-PI3K, p101 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23533 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction