Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIK3R5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562573
Description
Immunogen sequence: ATTCTEDRIQ HALERCLHGL SLSRRSTSWS AGLCLNCWSL QELVSRDPGH FLILLEQILQ KTREVQEKGT YDLLTPLALL FYSTVLCTPH FPPDSDLLLK AASTYH Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Receptor-regulated class I phosphoinositide 3-kinases.
Specifications
PIK3R5 | |
Polyclonal | |
Unconjugated | |
PIK3R5 | |
AV230647; F730038I15Rik; FOAP-2; p101; p101-PI3K; phosphatidylinositol 4,5-bisphosphate 3-kinase regulatory subunit; phosphatidylinositol-4,5-bisphosphate 3-kinase regulatory subunit; phosphoinositide 3-kinase regulatory subunit 5; phosphoinositide-3-kinase regulatory subunit 5; phosphoinositide-3-kinase, regulatory subunit 5; phosphoinositide-3-kinase, regulatory subunit 5, p101; PI3-kinase p101 subunit; PI3-kinase regulatory subunit 5; Pik3r5; Protein FOAP-2; ptdIns-3-kinase p101; PtdIns-3-kinase regulatory subunit; RGD1563261 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23533 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WYR1 | |
PIK3R5 | |
Recombinant protein corresponding to Human PIK3R5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction