Learn More
PIK3R5 Mouse anti-Canine, Human, Mouse, Non-human primate, Rat, Clone: OTI2A6, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA526211
Catalog No. PIMA526211
Add to cart
Description
Description
Receptor-regulated class I phosphoinositide 3-kinases (PI3Ks) phosphorylate the membrane lipid phosphatidylinositol 4,5-bisphosphate (PtdIns(4,5)P2) to PtdIns(3,4,5)P3, which in turn recruits and activates cytosolic effectors involved in proliferation, survival, or chemotaxis. PIK3R5 is a PI3K regulatory subunit (Brock et al., 2003 [PubMed 12507995]).
Specifications
Specifications
PIK3R5
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8WYR1, Q5SW28
Phosphoinositide 3-kinase regulatory subunit 5; PI3-kinase regulatory subunit 5; PI3-kinase p101 subunit; PtdIns-3-kinase p101; p101-PI3K; Phosphatidylinositol-4; 5-bisphosphate 3-kinase regulatory subunit; PtdIns-3-kinase regulatory subunit; Protein FOAP-2; PIK3R5;
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23533, 320207, 497931, 612805, 722027
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2A6
Unconjugated
PIK3R5
Liquid
Pik3r5
Full length human recombit protein of PIK3R5 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Mouse, Non-human primate, Rat