Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI4KB Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549603

Catalog No. PA549603


Add to cart

Description

Description

Predicted to react with bovine, zebrafish, mouse and rat based on sequence homology.

Phosphorylates phosphatidylinositol (PI) in the first committed step in the production of the second messenger inositol-1, 4, 5, -trisphosphate (PIP). PI4KB may regulate Golgi disintegration/reorganization during mitosis, possibly via its phosphorylation.
Specifications

Specifications

PI4KB
Polyclonal
PBS with 0.09% sodium azide
Q9UBF8
Phosphatidylinositol 4-kinase beta, PI4K-beta, PI4Kbeta, PtdIns 4-kinase beta, NPIK, PI4K92, PI4KB, PIK4CB
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5298
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PI4KB
Liquid
PI4KB
KLH conjugated synthetic peptide between 519-548 amino acids from the Central region of human PIK4CB.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit