Learn More
Abnova™ PIKFYVE (S696N/L932S/Q995L/T998S/S1033A/Q1183K) (Human) Recombinant Protein
For use with Functional Study.
Manufacturer:  Abnova™ P6570
Catalog No. 89-339-431
Add to cart
Description
Description
Human PIKFYVE (NP_055855.2, 1 a.a. - 2098 a.a.) S696N/L932S/Q995L/T998S/S1033A/Q1183K mutant full length recombinant protein with GST-tag at N-terminal using baculovirus expression system.
Specifications
Specifications
Functional Study
phosphatidylinositol-3-phosphate/phosphatidylinositol 5-kinase, type III
5 µg
CFD, FAB1, KIAA0981, MGC40423, PIKFYVE, PIP5K
200576
The purity was assessed by SDS-PAGE/CBB staining.
Stored at -80°C. Aliquot to avoid repeated freezing and thawing.